missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dermasel Agar

Isolate and identify dermatophyte fungi from hair, skin and nail samples with this selective agar.

Brand:  Thermo Scientific™ PO5037A

14.05 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10716223

  • 14.95€ / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Isolate and identify dermatophytes and other fungi from hair, nail or skin scrapings using selective Thermo Scientific™ Oxoid™ Dermasel Agar. Dermatophytes cultured on Dermasel Agar show characteristic colonial morphology with typical pigmentation. Macroconidia and microconidia are typical for the species when studied microscopically for identification.

Identify fungal scalp, skin and nail infections with Dermasel Agar, formulated for the primary isolation and identification of dermatophyte fungi.

  • Near neutral pH aids fungal growth
  • Cycloheximide inhibits growth of saprophytic fungi, yeasts and bacterial skin flora
  • Characteristic colonial morphology, typical pigmentation, macroconidia and microconidia aid identification
  • Ready-to-use convenience of prepared media

Emmons1 suggested that media for growth of dermatophytes should have a pH of 6.8 to 7.0 rather than pH 5.6 as is often recommended. A near neutral pH is better for the growth of some fungi, and the acid pH used to suppress bacterial contaminants can be replaced by antibiotics.

The addition to the medium of Dermasel Selective Supplement (Part No. SR0075E) to give a level of cycloheximide 0.4g/L and chloramphenicol 0.05g/L renders the medium selective for dermatophytes, inhibiting the growth of saprophytic fungi, yeasts and bacterial skin flora.

The addition of cycloheximide and an anti-bacterial agent has been reported to considerably improve the isolation of dermatophytes, especially when the inoculum, such as horse hair, was heavily contaminated 2,3. The presence of staphylococci, which may grow in the absence of the antibiotic, has been shown to prevent the in vitro growth of Trichophyton rubrum. The presence of cycloheximide in the medium inhibits the growth of Trichosporon cutaneum, Candida parasilosis, Candida krusei, Aspergillus, Penicillium, Fusarium and Cephalosporium species which have been associated with diseased nails.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 12°C, in the dark
90mm Monoplate
17.5g ± 5%
10/Pk.
Dermasel Agar
Agar
Wrapped in single cellulose-based film wrap
Product Suggestions

Product Suggestions

Videos
SDS
Documents Product Certifications

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6249148 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberPO5037A
Lot Number6240946 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberPO5037A
Lot Number6231235 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberPO5037A
Lot Number6218992 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberPO5037A
Lot Number6215661 Certificate TypeCertificate of Analysis Date02/01/2025 Catalog NumberPO5037A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Dermasel Agar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.