Learn More
Thermo Scientific™ Dermasel Agar
Isolate and identify dermatophyte fungi from hair, skin and nail samples with this selective agar.
15.35€
Specifications
Description | Dermasel Agar |
---|---|
Format | 90mm Monoplate |
Product Type | Agar |
Quantity | 10/Pk. |
Description
Isolate and identify dermatophytes and other fungi from hair, nail or skin scrapings using selective Thermo Scientific™ Oxoid™ Dermasel Agar. Dermatophytes cultured on Dermasel Agar show characteristic colonial morphology with typical pigmentation. Macroconidia and microconidia are typical for the species when studied microscopically for identification.
Identify fungal scalp, skin and nail infections with Dermasel Agar, formulated for the primary isolation and identification of dermatophyte fungi.
- Near neutral pH aids fungal growth
- Cycloheximide inhibits growth of saprophytic fungi, yeasts and bacterial skin flora
- Characteristic colonial morphology, typical pigmentation, macroconidia and microconidia aid identification
- Ready-to-use convenience of prepared media
Emmons1 suggested that media for growth of dermatophytes should have a pH of 6.8 to 7.0 rather than pH 5.6 as is often recommended. A near neutral pH is better for the growth of some fungi, and the acid pH used to suppress bacterial contaminants can be replaced by antibiotics.
The addition to the medium of Dermasel Selective Supplement (Part No. SR0075E) to give a level of cycloheximide 0.4g/L and chloramphenicol 0.05g/L renders the medium selective for dermatophytes, inhibiting the growth of saprophytic fungi, yeasts and bacterial skin flora.
The addition of cycloheximide and an anti-bacterial agent has been reported to considerably improve the isolation of dermatophytes, especially when the inoculum, such as horse hair, was heavily contaminated 2,3. The presence of staphylococci, which may grow in the absence of the antibiotic, has been shown to prevent the in vitro growth of Trichophyton rubrum. The presence of cycloheximide in the medium inhibits the growth of Trichosporon cutaneum, Candida parasilosis, Candida krusei, Aspergillus, Penicillium, Fusarium and Cephalosporium species which have been associated with diseased nails.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Dermasel Agar | |
90mm Monoplate | |
Agar | |
10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.