missing translation for 'onlineSavingsMsg'
Learn More
Diethyl (bromodifluoromethyl)phosphonate, 97%, Thermo Scientific Chemicals Product Code.: 11339355

Diethyl (bromodifluoromethyl)phosphonate, 97%, Thermo Scientific Chemicals

Product Code. 11339355
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11339355

Brand: Thermo Scientific Alfa Aesar L16715.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diethyl (bromodifluoromethyl)phosphonate is used as a reactant for the preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3), synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps and P-C bond cleavage on basic hydrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl (bromodifluoromethyl)phosphonate is used as a reactant for the preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3), synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps and P-C bond cleavage on basic hydrolysis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65094-22-6
Molecular Formula C5H10BrF2O3P
Molecular Weight (g/mol) 267.01
MDL Number MFCD00069118
InChI Key QRADKVYIJIAENZ-UHFFFAOYSA-N
Synonym diethyl bromodifluoromethyl phosphonate, bromodifluoromethyl diethylphosphonate, diethyl bromodifluoromethanephosphonate, diethyl bromodifluoromethylphosphonate, bromodifluoromethyl phosphonic acid diethyl ester, 1-bromo difluoro methyl-ethoxyphosphoryl oxyethane, phosphonic acid, bromodifluoromethyl-, diethyl ester, pubchem24183, acmc-20ai7pShow MoreShow Less
PubChem CID 144101
IUPAC Name 1-[[bromo(difluoro)methyl]-ethoxyphosphoryl]oxyethane
SMILES CCOP(=O)(OCC)C(F)(F)Br

Specifications

Density 1.503
Boiling Point 99°C to 102°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.416
Quantity 1 g
Beilstein 1944045
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 267.01
Percent Purity 97%
Chemical Name or Material Diethyl (bromodifluoromethyl)phosphonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL16Z024 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16715.03,L16715.14,L16715.06
Lot NumberI07Y028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16715.03,L16715.14,L16715.06
Lot NumberC12W059 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16715.03,L16715.14,L16715.06
Lot NumberA19U038 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16715.03,L16715.14,L16715.06
Lot NumberW02H032 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL16715.03,L16715.14,L16715.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl (bromodifluoromethyl)phosphonate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.