Learn More
Diethyl (bromodifluoromethyl)phosphonate, 97%
Description
Diethyl (bromodifluoromethyl)phosphonate is used as a reactant for the preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3), synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps and P-C bond cleavage on basic hydrolysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diethyl (bromodifluoromethyl)phosphonate is used as a reactant for the preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3), synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps and P-C bond cleavage on basic hydrolysis.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.

Specifications
Specifications
Density | 1.503 |
Boiling Point | 99°C to 102°C (16 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.416 |
Quantity | 5 g |
Beilstein | 1944045 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 267.01 |
Percent Purity | 97% |
Chemical Name or Material | Diethyl (bromodifluoromethyl)phosphonate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
L16Z024 | Certificate of Analysis | 08/12/2021 | L16715.03,L16715.14,L16715.06 |
I07Y028 | Certificate of Analysis | 08/12/2021 | L16715.03,L16715.14,L16715.06 |
C12W059 | Certificate of Analysis | 08/12/2021 | L16715.03,L16715.14,L16715.06 |
A19U038 | Certificate of Analysis | 08/12/2021 | L16715.03,L16715.14,L16715.06 |
W02H032 | Certificate of Analysis | 07/12/2021 | L16715.03,L16715.14,L16715.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.