Learn More
Afora™ Dimroth Condenser
Provides more effective cooling than conventional coil condensers. Afora™ Dimroth Condenser is a cold finger, coiled, fluid-cooled condenser.
Brand: Afora™ 1594/E
This item is not returnable.
View return policy
Description
- Internal double spiral
- Coolant inlet and outlet reside at the top of the condenser
- Often used with rotary evaporators
- Complies with UNE 400316
Specifications
UNE 400316 | |
293 mm | |
14/23 |
Exchange Surface: 250cm2 | |
200 mm | |
Condenser |