Learn More
Poulten Graf™ Dispensers Fortuna Polyfixes
Attain precision with Volac Smart™ Micropipette. Poulten Graf™ Volac Smart™ Variable Volume Micropipette are perfect for pipetting small amount of liquid samples in the laboratory.
Brand: Poulten Graf™ 101 07148
This item is not returnable.
View return policy
Description
- Coated PTFE-piston
- Fast and easy volume setting with additional fine adjust
- Fix adapter made of PP
- Valve block made of PP
Specifications
Dispensers Fortuna Polyfix | |
50 mL | |
0.025 kg | |
NS 12/21 |