Learn More
Poulten Graf™ Dispensers Fortuna Polyfixes
Attain precision with Volac Smart™ Micropipette. Poulten Graf™ Volac Smart™ Variable Volume Micropipette are perfect for pipetting small amount of liquid samples in the laboratory.
270.00€ - 298.00€
Specifications
Product Type Dispensers Fortuna Polyfix
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11762358
View Documents
Poulten Graf™
101 07148
50 mL
298.00€
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
11752358
View Documents
Poulten Graf™
101 07138
10 mL
270.00€
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
Description
Description
  • Coated PTFE-piston
  • Fast and easy volume setting with additional fine adjust
  • Fix adapter made of PP
  • Valve block made of PP
Specifications
Specifications
Dispensers Fortuna Polyfix