Learn More
Poulten Graf™ Dispensers Fortuna Polyfixes
Attain precision with Volac Smart™ Micropipette. Poulten Graf™ Volac Smart™ Variable Volume Micropipette are perfect for pipetting small amount of liquid samples in the laboratory.
270.00€ - 298.00€
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11762358
|
Poulten Graf™
101 07148 |
50 mL |
298.00€
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
11752358
|
Poulten Graf™
101 07138 |
10 mL |
270.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
Description
- Coated PTFE-piston
- Fast and easy volume setting with additional fine adjust
- Fix adapter made of PP
- Valve block made of PP
Specifications
Dispensers Fortuna Polyfix |