Learn More
Poulten Graf™ Dispensers Fortuna Polyfixes
Attain precision with Volac Smart™ Micropipette. Poulten Graf™ Volac Smart™ Variable Volume Micropipette are perfect for pipetting small amount of liquid samples in the laboratory.
Brand:  Poulten Graf™ 101 07148
 View more versions of this product
Product Code. 11762358
  • 298.00€ / Each
Estimated Shipment: 12-12-2024
Description
  • Coated PTFE-piston
  • Fast and easy volume setting with additional fine adjust
  • Fix adapter made of PP
  • Valve block made of PP
Specifications
Specifications
Dispensers Fortuna Polyfix
50 mL
0.025 kg
NS 12/21