Learn More
Thermo Scientific™ Domed PCR Caps, strips of 8
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Brand: Thermo Scientific™ AB0265
39.22 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifications
Clear | |
8 | |
Strips of 8 Domed Caps | |
Yes |
Polypropylene | |
250 strips | |
PCR Plate Cap Strip | |
96-well Plate |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now