Learn More
Wesbart™ Double Platform Test Tube Rack
Test Tube racks are an effective way of storing Falcon/ Universal tubes, Bijous, cryogenic vials or any size glass bottles.
Brand:  Wesbart™ DP1825
 View more versions of this product
Product Code. 15713368
  • 36.95€ / Each
Only null left
Estimated Shipment: 22-07-2025
Description
  • Racks are available in aluminum and stainless steel with the option of a hand grip
  • Test Tube racks can be designed to fit in horizontal cold store or be free standing
  • Different size test tubes racks are available to suit your requirements
  • Labels can be fitted to racks so they can be easily identified
Specifications
Specifications
Double Platform Test Tube Rack
Aluminum
110 mm
110 mm
Tubes (18 mm)
25
60 mm
18 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Wesbart™ Double Platform Test Tube Rack > Holds: Up to 25 tests tubes, 18 mm dia.