Learn More
Wesbart™ Double Platform Test Tube Rack
Test Tube racks are an effective way of storing Falcon/ Universal tubes, Bijous, cryogenic vials or any size glass bottles.
36.95€ - 82.60€
Specifications
Product Type Double Platform Test Tube Rack
Material Aluminum
View More Specs
Products 7
Product Code Brand Holds Positions Price Quantity & Availability  
Product Code Brand Holds Positions Price Quantity & Availability  
11781704
View Documents
Wesbart™
DP1450
Tubes (14 mm) 50
41.15€
Each
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
15815781
View Documents
Wesbart™
DP17145HG
Tubes (15 mL), Tubes (17 mm) 145
82.60€
Each
Estimated Shipment: 24-06-2025
to see stock.
Add to basket
15713368
View Documents
Wesbart™
DP1825
Tubes (18 mm) 25
36.95€
Each
Estimated Shipment: 17-06-2025
to see stock.
Add to basket
11711714
View Documents
Wesbart™
DP18100
Tubes (18 mm) 100
65.25€
Each
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
15723368
View Documents
Wesbart™
DP3025
Tubes (30 mm) 25
47.20€
Each
Estimated Shipment: 01-07-2025
to see stock.
Add to basket
11731714
View Documents
Wesbart™
DP2040
Tubes (20 mm) 40
43.20€
Each
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
15751589
View Documents
Wesbart™
DP3054HG
Tubes (5 mL) 54
76.60€
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
Description
Description
  • Racks are available in aluminum and stainless steel with the option of a hand grip
  • Test Tube racks can be designed to fit in horizontal cold store or be free standing
  • Different size test tubes racks are available to suit your requirements
  • Labels can be fitted to racks so they can be easily identified
Specifications
Specifications
Double Platform Test Tube Rack
Aluminum
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Wesbart™ Double Platform Test Tube Rack