missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DPTN
Potent and selective A3 adenosine receptor antagonist
231.00€ - 896.00€
Specifications
Chemical Name or Material | N-[4-(3,5-Dimethylphenyl)-5-(4-pyridinyl)-2-thiazolyl]-3-pyridinecarboxamide dihydrochloride |
---|---|
CAS | 325767-87-1 |
Target | Adenosine A3 R |
Molecular Formula | C22H18N4OS.2HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18739426
|
Tocris Bioscience™
7628/50 |
50 mg |
896.00€
50mg |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
18718296
|
Tocris Bioscience™
7628/10 |
10 mg |
231.00€
10mg |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Description
DPTN is a potent and selective A3 adenosine receptor antagonist (Ki values are in the range 0.36 - 1.65 nM (hA3AR) and 1.6 - 8.53 nM (rA3AR)). Reduces intrapulmonary pressure when combined with Dexamethasone (Cat. No. 1126).Specifications
N-[4-(3,5-Dimethylphenyl)-5-(4-pyridinyl)-2-thiazolyl]-3-pyridinecarboxamide dihydrochloride | |
325767-87-1 | |
Adenosine A3 R | |
C22H18N4OS.2HCl |
Safety and Handling
Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.