missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DRAQ9™
304.00€ - 954.00€
Specifications
Content And Storage | -20°C |
---|---|
For Use With (Application) | Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18060914
|
Novus Biologicals™
NBP2-81128-200UL |
200 μL |
304.00€
200µL |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
18247298
|
Novus Biologicals™
NBP2-81128-1000UL |
1000 μL |
954.00€
1000µL |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
Specifications
-20°C | |
Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title