Learn More
LABPLAS Dry Polyurethane Sani-Sponge
Synthetic polyurethane sponge for sampling surfaces.
Brand:  LABPLAS KSS-61805
 View more versions of this product
Product Code. 17950155
  • 646.00€ / Pack of 400
Estimated Shipment: 06-03-2025
Description
  • SANI-SPONGE
  • Dry sponge
  • Dimensions: 114 x 229 mm
  • Holds: 450 mL
Specifications
Specifications
Hygiene Sponge Sampling Kit
Sterile
Gloves