Learn More
LABPLAS Dry Polyurethane Sani-Sponge
Synthetic polyurethane sponge for sampling surfaces.
539.00€ - 646.00€
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17970155
|
LABPLAS
KSS-61800 |
None |
539.00€
Pack of 400 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
17950155
|
LABPLAS
KSS-61805 |
Gloves |
646.00€
Pack of 400 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
Description
- SANI-SPONGE
- Dry sponge
- Dimensions: 114 x 229 mm
- Holds: 450 mL
Specifications
Hygiene Sponge Sampling Kit | |
Sterile |