missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DSCR2 Recombinant Protein
344.00€ - 521.00€
Specifications
Accession Number | AAH03619 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 8624 |
Molecular Weight (g/mol) | 57.42 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182682
|
Abnova™
H00008624-P01.10ug |
10 μg |
344.00€
10µg |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
16192682
|
Abnova™
H00008624-P01.25ug |
25 μg |
521.00€
25µg |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH03619 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
57.42 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMG1 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
8624 | |
DSCR2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAATFFGEVVKAPCRAGTEDEEEEEEGRRETPEDREVRLQLARKREVRLLRRQTKTSLEVSLLEKYPCSKFIIAIGNNAVAFLSSFVMNSGVWEEVGCAKLWNEWCRTTDTTHLSSTEAFCVFYHLKSNPSVFLCQCSCYVAEDQQYQWLEKVFGSCPRKNMQITILTCRHVTDYKTSESTGSLPSPFLRALKTQNFKDSACCPLLEQPNIVHDLPAAVLSYCQVWKIPAILYLCYTDVMKLDLITVEAFKPILSTRSLKGLVKNIPQSTEILKKLMTTNEIQSNIYT | |
C21LRP/DSCR2/LRPC21/PAC1 | |
PSMG1 | |
Wheat Germ (in vitro) | |
GST |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title