Learn More
STEMCELL Technologies EasySep™ Direct Human Neutrophil Isolation Kit
Immunomagnetic isolation of neutrophils directly from whole blood
Brand:  STEMCELL Technologies 19666
Encompass
Product Code. 15805912
  • 1063.50€ / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Direct Human Neutrophil Isolation Kit isolates functional, highly purified neutrophils directly from human whole blood by immunomagnetic negative selection
  • No lysis, density gradient centrifugation or other processing steps are required and isolat​ed cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications
  • This product replaces the EasySep™ Human Neutrophil Isolation Kit (Catalog #19257) for even faster cell isolations
Specifications
Specifications
Granulocytes and Subsets
Cell Isolation
100 mL whole blood