Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood B Cell Positive Selection Kit
Column-free immunomagnetic positive selection of human B cells for HLA analysis
Brand: STEMCELL Technologies 17886
This item is not returnable.
View return policy
Description
- The EasySep™ HLA Chimerism Whole Blood B Cell Positive Selection Kit is designed to isolate B cells from fresh whole blood or buffy coat by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19, CD20 and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD19 antigen is present on mature B cells and B cell progenitors, but is lost upon maturation to plasma cells
- The antigen CD20 is expressed on all stages of B cell development except on pro B cells or plasma cells
- This product replaces the EasySep™ HLA Whole Blood B Cell Positive Selection Kit (Catalog #18184HLA) for even faster cell isolations
Specifications
B Cells | |
60 mL buffy coat or whole blood |