Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood B Cell Positive Selection Kit
Column-free immunomagnetic positive selection of human B cells for HLA analysis
Brand:  STEMCELL Technologies 17886
Encompass
Product Code. 17137801
  • 1146.00€ / 1 set
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ HLA Chimerism Whole Blood B Cell Positive Selection Kit is designed to isolate B cells from fresh whole blood or buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19, CD20 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD19 antigen is present on mature B cells and B cell progenitors, but is lost upon maturation to plasma cells
  • The antigen CD20 is expressed on all stages of B cell development except on pro B cells or plasma cells
  • This product replaces the EasySep™ HLA Whole Blood B Cell Positive Selection Kit (Catalog #18184HLA) for even faster cell isolations
Specifications
Specifications
B Cells
60 mL buffy coat or whole blood