Learn More
STEMCELL Technologies EasySep™ Mouse Biotin Positive Selection Kit II
Immunomagnetic positive selection of mouse cells labeled with biotinylated antibodies
Brand:  STEMCELL Technologies 17665
Encompass
Product Code. 17137731
  • 888.00€ / 1 set
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Mouse Biotin Positive Selection Kit II is designed to isolate cells that are labeled with biotinylated antibodies
  • Desired cells are targeted with antibody complexes recognizing biotin and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • This product replaces the EasySep™ Mouse Biotin Positive Selection Kit (Catalog #18556)
Specifications
Specifications
B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells
Cell Isolation
1 x 109 Cells