Learn More
STEMCELL Technologies EasySep™ Mouse Biotin Positive Selection Kit II
Immunomagnetic positive selection of mouse cells labeled with biotinylated antibodies
Brand: STEMCELL Technologies 17665
This item is not returnable.
View return policy
Description
- The EasySep™ Mouse Biotin Positive Selection Kit II is designed to isolate cells that are labeled with biotinylated antibodies
- Desired cells are targeted with antibody complexes recognizing biotin and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This product replaces the EasySep™ Mouse Biotin Positive Selection Kit (Catalog #18556)
Specifications
B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells | |
Cell Isolation | |
1 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.