Learn More
Sartorius Entris™ II Advanced Analytical Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand:  Sartorius BCA224I-1CEU
3857.00 EUR valid until 2025-12-31
Use promo code "24305" to get your promotional price.
 View more versions of this product
Product Code. 16635653
  • 4060.00€ / Each
Description
  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • For accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in real-time level support
  • Increases reliability of weighing results using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE Certificate
Verified
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤1.5 sec.
0.2 A
D-sub connector, 9-pin; USB UTL; USB Device
220 g
0.1 mg
Electronic Analytical Balance
100/240 V (±10%)
50/60 Hz
Videos
  • Sartorius Entris II Advanced Line application statistic
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Sartorius Entris II Advanced Line application statistic
    • Sartorius Entris II Advanced Line application peakhold
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Sartorius Entris II Advanced Line application peakhold
      • Sartorius Entris II Advanced Line application components
        Video Player is loading.
        Current Time 0:00
        Duration 1:19
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 1:19
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Sartorius Entris II Advanced Line application components
      • Sartorius Entris II Advanced Line application weighing
        Video Player is loading.
        Current Time 0:00
        Duration 0:35
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:35
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Sartorius Entris II Advanced Line application weighing
      • Sartorius Entris II Advanced Line application mixing
        Video Player is loading.
        Current Time 0:00
        Duration 1:21
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 1:21
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Sartorius Entris II Advanced Line application mixing
      • Sartorius Entris II Advanced Line application conversion
        Video Player is loading.
        Current Time 0:00
        Duration 0:54
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:54
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Sartorius Entris II Advanced Line application conversion
      • Sartorius Entris II Advanced Line application percentage weighing
        Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
          Sartorius Entris II Advanced Line application percentage weighing
        • Sartorius Entris II Advanced Line application counting
          Video Player is loading.
          Current Time 0:00
          Duration 1:08
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 1:08
          1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          Sartorius Entris II Advanced Line application counting
        • Sartorius Entris II Advanced Line application density determination
          Video Player is loading.
          Current Time 0:00
          Duration 0:00
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 0:00
          1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
            Sartorius Entris II Advanced Line application density determination
          • Sartorius Entris II Advanced Line application pipette smart test
            Video Player is loading.
            Current Time 0:00
            Duration 1:31
            Loaded: 0%
            Stream Type LIVE
            Remaining Time 1:31
            1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • default, selected
            Sartorius Entris II Advanced Line application pipette smart test
          SDS
          Documents
          Documents
          Brochures
          Product Guides
          Certificates
          View accessory products
          Promotions
          Promotions
          Product Content Correction
          Product Title
          Sartorius Entris™ II Advanced Analytical Balances with Country-Specific Verification > 220 g, Verified