Learn More
Sartorius Entris™ II Advanced Analytical Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
3580.00€ - 4550.00€
Specifications
Certifications/Compliance | CE Certificate |
---|---|
Connection Type | D-sub connector, 9-pin; USB UTL; USB Device |
Display Type | Intuitive Graphic Touch Technology |
Readability | 0.1 mg |
Material | Glass, PBT, Stainless Steel |
Product Code | Brand | Capacity | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Description | Price | Quantity & Availability | ||||
16685683
|
Sartorius
BCA64I-1CEU |
60 g | Verified |
3580.00€
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
16626243
|
Sartorius
BCA124I-1CEU |
120 g | Verified |
3680.00€
Each |
In Stock
Log in to see stock. |
||||
16635653
|
Sartorius
BCA224I-1CEU |
220 g | Verified |
3960.00€
Each |
In Stock
Log in to see stock. |
||||
16626253
|
Sartorius
BCA324I-1CEU |
320 g | Verified |
4550.00€
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
||||
Descrizione
- Entris™ II Advanced balances - Quality, Value, and Consistency
- For accurate weighing results that can be trusted time after time
- Simplifies balance leveling with built-in real-time level support
- Increases reliability of weighing results using integrated protection systems
- Controlled access to balance settings with user management
- Gapless documentation thanks to CalAuditTrail
Specifica
CE Certificate | |
Intuitive Graphic Touch Technology | |
Glass, PBT, Stainless Steel | |
≤1.5 sec. | |
0.2 A |
D-sub connector, 9-pin; USB UTL; USB Device | |
0.1 mg | |
Electronic Analytical Balance | |
100/240 V (±10%) | |
50/60 Hz |