missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ETHE1 Protein

Highly purified. Generating reliable and reproducible results.

528.00€ - 1390.00€

Specifications

For Use With (Application) SDS-PAGE
Purification Method SDS-PAGE
Source Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18426970
View Documents
Novus Biologicals™
NBP1-78881-0.5mg
0.5 mg
1390.00€
0.50mg
Please to purchase this item. Need a web account? Register with us today!
18382941
View Documents
Novus Biologicals™
NBP1-78881-100UG
100μg
528.00€
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 13-254 of Human ETHE1 The Recombinant Human ETHE1 Protein is derived from E. coli. The Recombinant Human ETHE1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

SDS-PAGE
Human
RUO
Unconjugated
SDS-PAGE
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
ETHE1
>90%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ ETHE1 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.