missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1H-indazole-5-carboxylate, 95%, Thermo Scientific™
92.70€ - 306.00€
Chemical Identifiers
CAS | 192944-51-7 |
---|---|
Molecular Formula | C10H10N2O2 |
Molecular Weight (g/mol) | 190.20 |
MDL Number | MFCD03426220 |
InChI Key | SKABXDPLIJIWLR-UHFFFAOYSA-N |
Synonym | 1h-indazole-5-carboxylic acid ethyl ester, 1h-indazole-5-carboxylic acid, ethyl ester, 5-ethoxycarbonyl-1h-indazole, 1h-indazole-5-carboxylicacidethylester, pubchem18041, 5-ethoxycarbonylindazole, acmc-1c4yw, ethyl indazole-5-carboxylate, ethyl 2h-indazole-5-carboxylate, indazole-5-carboxylic acid ethyl ester |
PubChem CID | 1501980 |
IUPAC Name | ethyl 1H-indazole-5-carboxylate |
SMILES | CCOC(=O)C1=CC=C2NN=CC2=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
192944-51-7 | |
190.20 | |
SKABXDPLIJIWLR-UHFFFAOYSA-N | |
1501980 | |
CCOC(=O)C1=CC=C2NN=CC2=C1 |
C10H10N2O2 | |
MFCD03426220 | |
1h-indazole-5-carboxylic acid ethyl ester, 1h-indazole-5-carboxylic acid, ethyl ester, 5-ethoxycarbonyl-1h-indazole, 1h-indazole-5-carboxylicacidethylester, pubchem18041, 5-ethoxycarbonylindazole, acmc-1c4yw, ethyl indazole-5-carboxylate, ethyl 2h-indazole-5-carboxylate, indazole-5-carboxylic acid ethyl ester | |
ethyl 1H-indazole-5-carboxylate |
Specifications
192944-51-7 | |
C10H10N2O2 | |
1h-indazole-5-carboxylic acid ethyl ester, 1h-indazole-5-carboxylic acid, ethyl ester, 5-ethoxycarbonyl-1h-indazole, 1h-indazole-5-carboxylicacidethylester, pubchem18041, 5-ethoxycarbonylindazole, acmc-1c4yw, ethyl indazole-5-carboxylate, ethyl 2h-indazole-5-carboxylate, indazole-5-carboxylic acid ethyl ester | |
CCOC(=O)C1=CC=C2NN=CC2=C1 | |
190.20 | |
190.2 | |
Ethyl 1H-indazole-5-carboxylate |
122°C to 124°C | |
MFCD03426220 | |
SKABXDPLIJIWLR-UHFFFAOYSA-N | |
ethyl 1H-indazole-5-carboxylate | |
1501980 | |
95% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title