Learn More
Falcon™ Clear Assay Plate Lid
For 384-well (PS) and polyvinyl chloride (PVC) plates
Brand: Falcon™ 353958
133.66 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Polystyrene lid for 384 well and 96 well Falcon™ microplates with 120μL total well capacity
- 6mm polystyrene, for 96 and 384 well microplates
- Non-sterile
Specifications
Polystyrene | |
Non-sterile | |
Microplate lid | |
96/384-well Plate, 120 μL Wells |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13