Learn More
Value Plastics Female Luer to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Brand:  Value Plastics 50120-63
 View more versions of this product
Product Code. 30053368
  • 21.31€ / Pack of 25
Estimated Shipment: 24-06-2025
Explore more special offers
Add to basket
Description
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
Non-DEHP;Non-Phthalate;RoHS
Nylon
Female Luer x Hose Barb
3/32 in.
25/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Value Plastics Female Luer to 200 Series Barb > 3/32 in.,Nylon
Always test the application to verify the performance of the interconnection.