Learn More
Value Plastics Female Luer to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
21.31€ - 30.83€
Product Code | Brand | Diameter (English) Inner | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Material | Price | Quantity & Availability | |||||
30053083
|
Value Plastics
50120-38 |
1/8 in. | Nylon |
21.31€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30053501
|
Value Plastics
50120-49 |
1/16 in. | Nylon |
21.31€
Pack of 25 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
30054140
|
Value Plastics
50120-67 |
1/16 in. | Polycarbonate |
25.74€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30054270
|
Value Plastics
50120-37 |
1/16 in. | Polypropylene |
27.20€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30054126
|
Value Plastics
50120-64 |
5/32 in. | Nylon |
24.14€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30053387
|
Value Plastics
50120-45 |
5/32 in. | Polypropylene |
30.83€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30053941
|
Value Plastics
50120-50 |
3/32 in. | Polycarbonate |
25.74€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30052979
|
Value Plastics
50120-59 |
1/8 in. | Polycarbonate |
25.74€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30053616
|
Value Plastics
50120-70 |
3/16 in. | Polypropylene |
30.38€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30054218
|
Value Plastics
50120-44 |
3/32 in. | Polypropylene |
27.20€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30054301
|
Value Plastics
50120-39 |
3/16 in. | Nylon |
24.14€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30054266
|
Value Plastics
50120-51 |
5/32 in. | Polycarbonate |
28.34€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30053368
|
Value Plastics
50120-63 |
3/32 in. | Nylon |
21.31€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30053483
|
Value Plastics
50120-55 |
1/18 in. | Polypropylene |
27.43€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
30054005
|
Value Plastics
50120-68 |
3/16 in. | Polycarbonate |
28.34€
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtimeSpecifications
25/Pk. | |
Female Luer x Hose Barb |
Always test the application to verify the performance of the interconnection.
Product Title
Value Plastics Female Luer to 200 Series Barb