Learn More
Value Plastics Female Luer to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Brand:  Value Plastics 50120-55
 View more versions of this product
Product Code. 30053483
  • 27.43€ / Pack of 25
Only null left
Estimated Shipment: 02-07-2025
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Polypropylene
Female Luer x Hose Barb
1/18 in.
25/Pk.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Value Plastics Female Luer to 200 Series Barb > 1/18 in.,Polypropylene
Always test the application to verify the performance of the interconnection.