Learn More
Value Plastics Female Luer to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Brand:  Value Plastics 50120-45
 View more versions of this product
Product Code. 30053387
  • 30.83€ / Pack of 25
Estimated Shipment: 24-06-2025
Explore more special offers
Add to basket
Description
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Polypropylene
Female Luer x Hose Barb
5/32 in.
25/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Value Plastics Female Luer to 200 Series Barb > 5/32 in.,Polypropylene
Always test the application to verify the performance of the interconnection.