Learn More
Value Plastics Female Luer to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Brand:  Value Plastics 50120-39
 View more versions of this product
Product Code. 30054301
  • 24.14€ / Pack of 25
Estimated Shipment: 24-06-2025
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
Non-DEHP;Non-Phthalate;RoHS
Nylon
Female Luer x Hose Barb
3/16 in.
25/Pk.
Product Title
Value Plastics Female Luer to 200 Series Barb > 3/16 in.,Nylon
Always test the application to verify the performance of the interconnection.