missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FGF16 (Human) Recombinant Protein

Human FGF16 full-length ORF ( NP_003859.1, 1 a.a. - 207 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00008823-P01.10ug

 View more versions of this product

Product Code. 16182942

  • 344.00€ / 10µg
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sequence: MAEVGGVFASLDWDLHGFSSSLGNVPLADSPGFLNERLGQIEGKLQRGSPTDFAHLKGILRRRQLYCRTGFHLEIFPNGTVHGTRHDHSRFGILEFISLAVGLISIRGVDSGLYLGMNERGELYGSKKLTRECVFREQFEENWYNTYASTLYKHSDSERQYYVALNKDGSPREGYRTKRHQKFTHFLPRPVDPSKLPSMSRDLFHYR
Specifications

Specifications

NP_003859.1
fibroblast growth factor 16
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
8823
Wheat germ expression system
10 μg
FGF16
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.