Learn More
Witeg™ Gas Washing Bottles, Drechsel
Finely crafted gas washing bottle, according to Dreschel. Witeg™ Gas Washing Bottles, Drechsel does not include a sintered disc.
Brand: Witeg™ 2 400 001
This item is not returnable.
View return policy
Description
- For gas washing
- Without sintered dis
Specifications
1000 mL | |
Drechsel | |
Gas Washing Bottle | |
ST 29/32 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Witeg™ Gas Washing Bottles, Drechsel > Capacity: 1000mL