missing translation for 'onlineSavingsMsg'
Learn More

GATA-6 Antibody (SR2056), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-21806-100ul

 View more versions of this product

Product Code. 18681975

  • 448.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GATA-6 Monoclonal antibody specifically detects GATA-6 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GATA-6
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunofluorescence
SR2056
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
GATA binding protein 6, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6
A synthesized peptide derived from human GATA-6 (Uniprot #: Q92908)
100 μg
Phospho Specific, Stem Cell Markers
2627
Store at -20° C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
GATA-6 Antibody (SR2056), Novus Biologicals™ > 100 μg; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.