missing translation for 'onlineSavingsMsg'
Learn More

GATA-6 Antibody (SR2056), Novus Biologicals™

Rabbit Monoclonal Antibody

340.00€ - 448.00€

Specifications

Antigen GATA-6
Clone SR2056
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18681975
View Documents
Novus Biologicals
NBP3-21806-100ul
100 μg
448.00€
100µL
Please to purchase this item. Need a web account? Register with us today!
18630884
View Documents
Novus Biologicals
NBP3-21806-50ul
50 μg
340.00€
50µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GATA-6 Monoclonal antibody specifically detects GATA-6 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

GATA-6
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human
GATA binding protein 6, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6
A synthesized peptide derived from human GATA-6 (Uniprot #: Q92908)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2056
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Phospho Specific, Stem Cell Markers
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
2627
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
GATA-6 Antibody (SR2056), Novus Biologicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.