Learn More
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Brand:  Thermo Scientific™ 29001318
16.40 EUR valid until 2025-04-30
Use promo code "24211" to get your promotional price.
 View more versions of this product
Product Code. 13415099
  • 18.99€ / Each
In Stock
Description
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Liner Sealing Ring
1 Ea.
Liner Sealing Ring
TRACE 1300 Series GC PTV Injectors
Special Offers
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
    Days
    57
    Hours
    9
    Minutes
    16
    Seconds
    53
  • Multiple Manufacturers
    Discover exclusive monthly deals on chromatography supplies to boost your lab's efficiency.
    Promo Code: 24211
    Expires2025-04-30
    Days
    26
    Hours
    9
    Minutes
    16
    Seconds
    53
Product Content Correction
Product Title
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments >