Learn More
Jenway™ Genova-Series Spectrophotometer Accessory, Microcuvette Holder
Parts and accessories to expand analysis capabilities or make testing easier.
Brand: Jenway™ 83056-06
This item is not returnable.
View return policy
Description
- Accessories
- For use with Genova-Series Spectrophotometer
Specifications
10 x 10 mm square, 8.5 mm beam height | |
Microcuvette Holder | |
Genova-Series Spectrophotometer |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Jenway™ Genova-Series Spectrophotometer Accessory, Microcuvette Holder > Microcuvette Holder