Learn More
Rogo Sampaic™ Glass Slides
Brand:  Rogo Sampaic™ BPB029
 View more versions of this product
Product Code. 11822350
  • 9.65€ / Pack of 50
Estimated Shipment: 15-10-2024
Specifications
76 mm
Cut
Yellow
76 x 26 mm
26 mm
Glass
50 Pack