Learn More
Rogo Sampaic™ Class B Glass Volumetric Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Rogo Sampaic™ Class B Glass Volumetric Pipettes are manufactured from clear soda lime glass.
Brand: Rogo Sampaic™ DGG004
This item is not returnable.
View return policy
Description
- With vitrified amber graduations
- With safety bulb
Specifications
Yes | |
Glass | |
0.1 mL | |
10 mL | |
Graduated Pipette |
B | |
Yes | |
1 Pack | |
360 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Product Title
Rogo Sampaic™ Class B Glass Volumetric Pipettes > Capacity: 10mL