missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen SA510280
This item is not returnable.
View return policy
Description
Canine IgG (H+L) Polyclonal antibody specifically detects Canine IgG (H+L) in Canine samples. It is validated for ELISA, Flow Cytometry, Immunoelectrophoresis, Immunohistochemistry, Western Blot
Specifications
Canine IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Canine | |
Liquid |
ELISA, Flow Cytometry, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Canine IgG h+l | |
1 mg | |
Secondary | |
2-8°C | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Goat anti-Canine IgG (H+L) Secondary Antibody, Invitrogen™ > 1 mg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction