Learn More
Abnova™ GPCR Signaling to MAPK/ERk Phospho-Specific Array
GPCR Signaling to MAPK/ERk Phospho-Specific Array includes 193 highly specific and well-characterized phosphorylation antibodies related to this signaling pathway.
Brand:  Abnova™ AA0027.1Kit(2Samples)
Product Code. 16052435
  • 1125.00€ / 1 set
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • This product must be accompanied with Antibody Array Detection Kit
  • The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
  • Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum
Specifications
Specifications
4°C
1 Kit (2 Samples)
Array/Library
Fluorometric
Profiling