Learn More
Hirschmann™ Graduated Pipettes with Piston
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Graduated Pipettes with Piston come with amber stain graduation.
Brand: Hirschmann™ 1190170
This item is not returnable.
View return policy
Description
- Graduated to tip
- Available in a range of capacities
Specifications
Yes | |
0.1 mL | |
1 Each | |
475 mm |
Amber | |
Amber graduations permanently with silver diffusion color | |
25 mL | |
Graduated Pipet |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Hirschmann™ Graduated Pipettes with Piston > Capacity: 25mL; Length: 475mm