missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HDCMA18P Recombinant Protein
Brand: Abnova™ H00051574-P01.10ug
This item is not returnable.
View return policy
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH06981 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
29.15 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
METESGNQEKVMEEESTEKKKEVEKKETVTS | |
DKFZp564K112/HDCMA18P/MGC104360/PIP7S | |
LARP7 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
51574 | |
HDCMA18P (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LARP7 | |
Human | |
Recombinant | |
Solution |