missing translation for 'onlineSavingsMsg'
Learn More

Scotch-Brite™ High Performance Cleaning Wipes 2011

Long lasting, high performance cleaning wipes. Can be used to dry or damp for a lint-free, streak-free finish.

Brand:  Scotch-Brite™ 7100006244

 View more versions of this product

Product Code. 17118680

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For removal of dust, dirt, grease, and fingerprints
  • Use on stainless steel, chrome, plastics, glass, paint lacquers, and other finished or polished metal surfaces
  • Eliminates the need for cleaning agents in many cases
  • Machine washable up to 75 times at 95°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wipes
320 mm
360 x 320 mm
Machine Washable
360 mm
Red
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers