missing translation for 'onlineSavingsMsg'
Learn More

Scotch-Brite™ High Performance Cleaning Wipes 2011

Long lasting, high performance cleaning wipes. Can be used to dry or damp for a lint-free, streak-free finish.

Brand:  Scotch-Brite™ 7100010928

 View more versions of this product

Product Code. 17168680

  • 7095.00€ / Pack of 400
Estimated Shipment: 21-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For removal of dust, dirt, grease, and fingerprints
  • Use on stainless steel, chrome, plastics, glass, paint lacquers, and other finished or polished metal surfaces
  • Eliminates the need for cleaning agents in many cases
  • Machine washable up to 75 times at 95°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cleaning Wipe
320 mm
360 x 320 mm
Machine Washable
360 mm
Blue
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers