Learn More
Abnova™ HotTaq DNA Polymerase
HotTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
Brand:  Abnova™ D0011.500Units
 View more versions of this product
Product Code. 16012555
  • 262.00€ / 500 units
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The enzyme has been proved not having the 3′ - 5′exonuclease activity.
  • Prior the first PCR step the HotTaq DNA polymerase should be activated by 15 minute incubation at 95-97°C.
Specifications
Specifications
Store at -20°C Guaranteed stable for 6 months when properly stored
Standard PCR
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
500 units
10 units/μL