missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HotTaq DNA Polymerase

HotTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.

262.00€ - 1010.00€

Specifications

Content And Storage Store at -20°C Guaranteed stable for 6 months when properly stored
For Use With (Application) Standard PCR
Concentration 10 units/ÎĽL
Storage Buffer 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16002555
View Documents
Abnova™
D0011.2500Units
2500 units
1010.00€
2500 units
Please to purchase this item. Need a web account? Register with us today!
16012555
View Documents
Abnova™
D0011.500Units
500 units
262.00€
500 units
Please to purchase this item. Need a web account? Register with us today!
16092545
View Documents
Abnova™
D0011.1000Units
1000 units
508.00€
1000 units
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The enzyme has been proved not having the 3′ - 5′exonuclease activity.
  • Prior the first PCR step the HotTaq DNA polymerase should be activated by 15 minute incubation at 95-97°C.
Specifications

Specifications

Store at -20°C Guaranteed stable for 6 months when properly stored
Standard PCR
10 units/ÎĽL
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ HotTaq DNA Polymerase

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.