Learn More
ACROBiosystems Human CD30 / TNFRSF8 Protein, His Tag
339.67€ - 2181.73€
Specifications
Accession Number | NP_001234.2 |
---|---|
Molecular Weight (g/mol) | 40.4 kDa |
Source | Human CD30, His Tag (CD0-H52H5) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). |
Immunogen | Phe 19 - Lys 379 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17206080
|
ACROBiosystems
CD0-H52H5-1mg |
1 mg |
2181.73€
1mg |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
17296070
|
ACROBiosystems
CD0-H52H5-100ug |
100 μg |
339.67€
100µg |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
Description
Human CD30 is also known as TNFRSF8, is a cell membrane protein of the tumor necrosis factor receptor family and tumor marker. TNFRSF-8 is expressed by activated, but not by resting, T and B cells. Also, CD30 is expressed on classical Hodgkin Lymphoma cells together with CD15. CD30 is the receptor for TNFSF8/CD30L. CD30 can interact with TRAF2 and TRAF5, and mediate the signal transduction that leads to the activation of NF-kappa-B. TNFRSF8 may play a role in the regulation of cellular growth and transformation of activated lymphoblasts. TNFRSF8 is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.4 kDa.Specifications
NP_001234.2 | |
Human CD30, His Tag (CD0-H52H5) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). | |
-20°C | |
CD30 | |
Human | |
HEK293 | |
Immune Checkpoint Proteins |
40.4 kDa | |
Phe 19 - Lys 379 | |
TNFRSF8,CD30,D1S166E,Ki-1 | |
Unconjugated | |
His Tag | |
Powder |