Learn More
ACROBiosystems Human CLEC2D Protein, Mouse IgG2a Fc Tag
Brand: ACROBiosystems CLD-H5253-1mg
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
CLEC2D (C-type lectin domain family 2 member D) is also known as CLAX, LLT1, OCIL is receptor for KLRB1 that protects target cells against natural killer cell-mediated lysis, inhibits osteoclast formation, inhibits bone resorption and modulates the release of interferon-gamma. It is also a ligand for the human NKR-P1A (CD161) receptor, present on NK cells and T cells. Furthermore, The reference shows the pathway may as a potential immunotherapy targets.
This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 42.2 kDa.Specifications
42.2 kDa | |
Human CLEC2D, Mouse IgG2a Fc Tag (CLD-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 60 - Val 191 (Accession # Q9UHP7-1). | |
-20°C | |
CLEC2D | |
Human | |
HEK293 | |
Bio-Markers & CD Antigens |
1 mg | |
Arg 60 - Val 191 | |
CLEC2D, C-type lectin domain family 2 member D, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT-1, Osteoclast inhibitory lectin | |
Unconjugated | |
Mouse IgG Fc Tag | |
Powder |