missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ENPP7 Full-length ORF (AAH41453.2, 1 a.a. - 458 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00339221-P01.25ug
This item is not returnable.
View return policy
Description
Sequence: MRGPAVLLTVALATLLAPGAGAPVQSQGSQNKLLLVSFDGFRWNYDQDVDTPNLDAMARDGVKARYMTPAFVTMTSPCHFTLVTGKYIENHGVVHNMYYNTTSKVKLPYHATLGIQRWWDNGSVPIWITAQRQGLRAGSFFYPGGNVTYQGVAVTRSRKEGIAHNYKNETEWRANIDTVMAWFTEEDLDLVTLYFGEPDSTGHRYGPESPERREMVRQVDRTVGYLRESIARNHLTDRLNLIITSDHGMTTVDKRAGDLVEFHKFPNFTFRDIEFELLDYGPNGMLLPKEGRLEKVYDALKDAHPKLHVYKKEAFPEAFHYANNPRVTPLLMYSDLGYVIHGRINVQFNNGEHGFDNKDMDMKTIFRAVGPSFRAGLEVEPFESVHVYELMCRLLGIVPEANDGHLATLLPMLHTESALPPDGRPTLLPKGRSALPPSSRPLLVMGLLGTVILLSEVASpecifications
AAH41453.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALK-SMase/MGC50179 | |
ENPP7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
339221 | |
ENPP7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRGPAVLLTVALATLLAPGAGAPVQSQGSQNKLLLVSFDGFRWNYDQDVDTPNLDAMARDGVKARYMTPAFVTMTSPCHFTLVTGKYIENHGVVHNMYYNTTSKVKLPYHATLGIQRWWDNGSVPIWITAQRQGLRAGSFFYPGGNVTYQGVAVTRSRKEGIAHNYKNETEWRANIDTVMAWFTEEDLDLVTLYFGEPDSTGHRYGPESPERREMVRQVDRTVGYLRESIARNHLTDRLNLIITSDHGMTTVDKRAGDLVEFHKFPNFTFRDIEFELLDYGPNGMLLPKEGRLEKVYDALKDAHPKLHVYKKEAFPEAFHYANNPRVTPLLMYSDLGYVIHGRINVQFNNGEHGFDNKDMDMKTIFRAVGPSFRAGLEVEPFESVHVYELMCRLLGIVPEANDGHLATLLPMLHTESALPPDGRPTLLPKGRSALPPSSRPLLVMGLLGTVILLSEVA | |
RUO | |
ENPP7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |