missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FLJ21019 Partial ORF (NP_079203, 665 a.a. - 750 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079990-Q01.10ug
This item is not returnable.
View return policy
Description
Sequence: DVLELSTEPGRGAPQKLCLGLGAKAMSLSRPGETEPIHSVSYGHVAACQLMGPHTLALRVGESQLLLQSPQVEEIMQLVNAYLANPSpecifications
NP_079203 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.2kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DVLELSTEPGRGAPQKLCLGLGAKAMSLSRPGETEPIHSVSYGHVAACQLMGPHTLALRVGESQLLLQSPQVEEIMQLVNAYLANP | |
RUO | |
PLEKHH3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
79990 | |
FLJ21019 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ21019 | |
PLEKHH3 | |
Recombinant | |
wheat germ expression system |