Learn More
ACROBiosystems Human HGF Protein, Fc Tag
Brand:  ACROBiosystems HGF-H5253-1mg
Encompass_Preferred
 View more versions of this product
Product Code. 17216230
  • 3775.58€ / 1mg
Estimated Shipment: 25-09-2024
Description
Hepatocyte growth factor (HGF) is a paracrine cellular growth, motility and morphogenic factor. Activating ligand for the receptor tyrosine kinase MET by binding to it and promoting its dimerization. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. In addition, HGF has been implicated in a variety of cancers, including of the lungs, pancreas, thyroid, colon, and breast.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 106.1 kDa | 53.7 kDa | 52.4 kDa.
Specifications
Specifications
106.1 kDa, 53.7 kDa, 52.4 kDa
Human HGF, Fc Tag (HGF-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Ser 728 (Accession # P14210-1).
-20°C
HGF
Human
HEK293
Cytokines&Growth; Factors
1 mg
Gln 32 - Ser 728
HGF,HPTA,SF
Unconjugated
Fc Tag
Powder