Learn More
ACROBiosystems Human HGF Protein, Fc Tag
Brand: ACROBiosystems HGF-H5253-1mg
This item is not returnable.
View return policy
Description
Hepatocyte growth factor (HGF) is a paracrine cellular growth, motility and morphogenic factor. Activating ligand for the receptor tyrosine kinase MET by binding to it and promoting its dimerization. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. In addition, HGF has been implicated in a variety of cancers, including of the lungs, pancreas, thyroid, colon, and breast.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 106.1 kDa | 53.7 kDa | 52.4 kDa.Specifications
106.1 kDa, 53.7 kDa, 52.4 kDa | |
Human HGF, Fc Tag (HGF-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Ser 728 (Accession # P14210-1). | |
-20°C | |
HGF | |
Human | |
HEK293 | |
Cytokines&Growth; Factors |
1 mg | |
Gln 32 - Ser 728 | |
HGF,HPTA,SF | |
Unconjugated | |
Fc Tag | |
Powder |